Product Details

SNP ID
rs200866589
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:722313 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGCTGTTCTCTCCCTCAGCTCC[C/G]GCTGCTGGAGGACAAGCTGCGGACA
Phenotype
MIM: 614666 MIM: 610998 MIM: 611118
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CCDC78 PubMed Links

Gene Details

Gene
CCDC78
Gene Name
coiled-coil domain containing 78
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031737.2 597 Intron NP_001026907.2
XM_006720838.1 597 Intron XP_006720901.1
XM_006720843.3 597 Intron XP_006720906.1
XM_011522356.1 597 Intron XP_011520658.1
XM_011522357.1 597 Intron XP_011520659.1
XM_011522358.2 597 Intron XP_011520660.1
XM_011522359.1 597 Intron XP_011520661.1
XM_011522360.1 597 Intron XP_011520662.1
XM_011522361.1 597 Intron XP_011520663.1
XM_011522362.1 597 Intron XP_011520664.1
XM_011522363.1 597 Intron XP_011520665.1
XM_011522364.1 597 Intron XP_011520666.1
XM_011522365.1 597 Intron XP_011520667.1
XM_011522366.1 597 Intron XP_011520668.1
XM_011522367.1 597 Intron XP_011520669.1
XM_011522368.1 597 Intron XP_011520670.1
XM_011522369.1 597 Intron XP_011520671.1
XM_011522370.1 597 Intron XP_011520672.1
XM_011522371.2 597 Intron XP_011520673.1
XM_017022929.1 597 Intron XP_016878418.1
XM_017022930.1 597 Intron XP_016878419.1
XM_017022931.1 597 Intron XP_016878420.1
XM_017022932.1 597 Intron XP_016878421.1
Gene
FAM173A
Gene Name
family with sequence similarity 173 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271285.1 597 Missense Mutation CCG,CGG P138R NP_001258214.1
NM_023933.2 597 Missense Mutation CCG,CGG P155R NP_076422.1
Gene
HAGHL
Gene Name
hydroxyacylglutathione hydrolase-like
There are no transcripts associated with this gene.

Gene
METRN
Gene Name
meteorin, glial cell differentiation regulator
There are no transcripts associated with this gene.

Gene
NARFL
Gene Name
nuclear prelamin A recognition factor like
There are no transcripts associated with this gene.

View Full Product Details