Product Details
- SNP ID
-
rs199704165
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:70810006 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGCTGAAATTCAAATGAGAACGTG[C/T]CCTGGAAGAGAAAACAGAGGATCCT
- Phenotype
-
MIM: 610812
MIM: 604632
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
HYDIN
PubMed Links
Gene Details
- Gene
- HYDIN
- Gene Name
- HYDIN, axonemal central pair apparatus protein
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001198542.1 |
14873 |
Intron |
|
|
NP_001185471.1 |
NM_001198543.1 |
14873 |
Intron |
|
|
NP_001185472.1 |
NM_001270974.2 |
14873 |
Missense Mutation |
GAC,GGC |
D4887G |
NP_001257903.1 |
NM_017558.4 |
14873 |
Intron |
|
|
NP_060028.2 |
XM_006721206.2 |
14873 |
Missense Mutation |
GAC,GGC |
D4904G |
XP_006721269.1 |
XM_011523146.2 |
14873 |
Missense Mutation |
GAC,GGC |
D4948G |
XP_011521448.1 |
XM_011523147.1 |
14873 |
Missense Mutation |
GAC,GGC |
D4938G |
XP_011521449.1 |
XM_011523148.1 |
14873 |
Missense Mutation |
GAC,GGC |
D4921G |
XP_011521450.1 |
XM_011523151.1 |
14873 |
Missense Mutation |
GAC,GGC |
D4914G |
XP_011521453.1 |
XM_011523152.1 |
14873 |
Missense Mutation |
GAC,GGC |
D2841G |
XP_011521454.1 |
XM_011523155.2 |
14873 |
Missense Mutation |
GAC,GGC |
D2517G |
XP_011521457.1 |
XM_017023346.1 |
14873 |
Missense Mutation |
GAC,GGC |
D4927G |
XP_016878835.1 |
XM_017023347.1 |
14873 |
Missense Mutation |
GAC,GGC |
D4291G |
XP_016878836.1 |
XM_017023348.1 |
14873 |
Missense Mutation |
GAC,GGC |
D4291G |
XP_016878837.1 |
- Gene
- VAC14
- Gene Name
- Vac14, PIKFYVE complex component
There are no transcripts associated with this gene.
View Full Product Details