Product Details

SNP ID
rs201927760
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:786194 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGCTCCAGCAGGCATCCAGGGAG[A/G]GCAGGAAGGGGTCAGGCGTGCAGAC
Phenotype
MIM: 613201
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CHTF18 PubMed Links
Additional Information
For this assay, SNP(s) [rs3751672] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
CHTF18
Gene Name
chromosome transmission fidelity factor 18
There are no transcripts associated with this gene.

Gene
RPUSD1
Gene Name
RNA pseudouridylate synthase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324086.1 740 Missense Mutation CCC,CTC P235L NP_001311015.1
NM_001324410.1 740 Missense Mutation CCC,CTC P106L NP_001311339.1
NM_001324411.1 740 UTR 3 NP_001311340.1
NM_001324412.1 740 Missense Mutation CCC,CTC P103L NP_001311341.1
NM_001324413.1 740 Missense Mutation CCC,CTC P103L NP_001311342.1
NM_001324414.1 740 Missense Mutation CCC,CTC P103L NP_001311343.1
NM_001324415.1 740 Missense Mutation CCC,CTC P103L NP_001311344.1
NM_058192.2 740 Missense Mutation CCC,CTC P232L NP_478072.1

View Full Product Details