Product Details

SNP ID
rs200240496
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:31318616 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATAATTAAGCAAATTAGCATAGCC[A/G]TGTTGTTGTTGTTTTCCGCACAGAC
Phenotype
MIM: 158380 MIM: 158381 MIM: 613113
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
EVI2A PubMed Links

Gene Details

Gene
EVI2A
Gene Name
ecotropic viral integration site 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003927.2 663 Missense Mutation ACG,ATG T156M NP_001003927.1
NM_014210.3 663 Missense Mutation ACG,ATG T133M NP_055025.2
Gene
EVI2B
Gene Name
ecotropic viral integration site 2B
There are no transcripts associated with this gene.

Gene
NF1
Gene Name
neurofibromin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000267.3 663 Intron NP_000258.1
NM_001042492.2 663 Intron NP_001035957.1
NM_001128147.2 663 Intron NP_001121619.1
XM_005257983.2 663 Intron XP_005258040.1
XM_005257984.2 663 Intron XP_005258041.1
XM_006721922.2 663 Intron XP_006721985.1
XM_006721924.2 663 Intron XP_006721987.1
XM_006721925.2 663 Intron XP_006721988.1
XM_006721926.3 663 Intron XP_006721989.1
XM_011524852.2 663 Intron XP_011523154.1
XM_011524857.2 663 Intron XP_011523159.1
XM_017024689.1 663 Intron XP_016880178.1
XM_017024690.1 663 Intron XP_016880179.1

View Full Product Details