Product Details

SNP ID
rs199812075
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:63881029 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTGGGGAGAAGGCATCCACTCAC[A/G]GATTTCTGCTGCGTTTTCACCCTGT
Phenotype
MIM: 118820 MIM: 139240
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CSH2 PubMed Links

Gene Details

Gene
CSH2
Gene Name
chorionic somatomammotropin hormone 2
There are no transcripts associated with this gene.

Gene
GH2
Gene Name
growth hormone 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002059.4 432 Silent Mutation TCC,TCT S97S NP_002050.1
NM_022556.3 432 Silent Mutation TCC,TCT S82S NP_072050.1
NM_022557.3 432 Silent Mutation TCC,TCT S97S NP_072051.1
NM_022558.3 432 Silent Mutation TCC,TCT S97S NP_072052.1
Gene
LOC107987248
Gene Name
intercellular adhesion molecule 1-like
There are no transcripts associated with this gene.

View Full Product Details