Product Details

SNP ID
rs200392888
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:19909229 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCATACTGAGCCTGCTGCATAAT[A/G]TCACTGACTATCATTTTAGCAATCT
Phenotype
MIM: 604694
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
AKAP10 PubMed Links
Additional Information
For this assay, SNP(s) [rs203462] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
AKAP10
Gene Name
A-kinase anchoring protein 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007202.3 2112 Silent Mutation GAC,GAT D645D NP_009133.2
XM_006721432.3 2112 Silent Mutation GAC,GAT D587D XP_006721495.1
XM_017024071.1 2112 Intron XP_016879560.1

View Full Product Details