Product Details

SNP ID
rs201887461
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:75501116 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCGTCCAGCTGGTCAGCCAGGGC[A/G]TCGAACATGGTGCTGATGTCATCCA
Phenotype
MIM: 612185
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CASKIN2 PubMed Links

Gene Details

Gene
CASKIN2
Gene Name
CASK interacting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142643.2 4160 Silent Mutation GAC,GAT D1109D NP_001136115.1
NM_020753.4 4160 Silent Mutation GAC,GAT D1191D NP_065804.2
Gene
MIR6785
Gene Name
microRNA 6785
There are no transcripts associated with this gene.

Gene
TMEM94
Gene Name
transmembrane protein 94
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321148.1 4160 Intron NP_001308077.1
NM_001321149.1 4160 Intron NP_001308078.1
NM_014738.5 4160 Intron NP_055553.3
XM_005257864.2 4160 Intron XP_005257921.1
XM_011525514.1 4160 Intron XP_011523816.1
XM_011525515.1 4160 Intron XP_011523817.1
XM_011525516.1 4160 Intron XP_011523818.1
XM_011525517.1 4160 Intron XP_011523819.1
XM_011525519.1 4160 Intron XP_011523821.1
XM_011525520.2 4160 Intron XP_011523822.1
XM_011525521.1 4160 Intron XP_011523823.1
XM_017025433.1 4160 Intron XP_016880922.1
XM_017025434.1 4160 Intron XP_016880923.1
XM_017025435.1 4160 Intron XP_016880924.1
XM_017025436.1 4160 Intron XP_016880925.1
XM_017025437.1 4160 Intron XP_016880926.1
XM_017025438.1 4160 Intron XP_016880927.1
XM_017025439.1 4160 Intron XP_016880928.1
XM_017025440.1 4160 Intron XP_016880929.1
XM_017025441.1 4160 Intron XP_016880930.1
XM_017025442.1 4160 Intron XP_016880931.1
XM_017025443.1 4160 Intron XP_016880932.1
XM_017025444.1 4160 Intron XP_016880933.1
XM_017025445.1 4160 Intron XP_016880934.1
XM_017025446.1 4160 Intron XP_016880935.1
XM_017025447.1 4160 Intron XP_016880936.1
XM_017025448.1 4160 Intron XP_016880937.1
XM_017025449.1 4160 Intron XP_016880938.1
XM_017025450.1 4160 Intron XP_016880939.1

View Full Product Details