Product Details

SNP ID
rs202240168
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:35855289 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTGGGTTCTCAGGCTCTTTCTGG[A/G]ACCTTGTGGGCTCTGCCTTCACATT
Phenotype
MIM: 601574
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HEATR9 PubMed Links

Gene Details

Gene
HEATR9
Gene Name
HEAT repeat containing 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321395.1 1579 Missense Mutation TCC,TTC S456F NP_001308324.1
NM_152781.3 1579 Missense Mutation TCC,TTC S496F NP_689994.2
XM_005278233.2 1579 Missense Mutation TCC,TTC S462F XP_005278290.1
XM_005278235.1 1579 Missense Mutation TCC,TTC S422F XP_005278292.1
XM_006721802.3 1579 Missense Mutation TCC,TTC S314F XP_006721865.1
XM_011524586.2 1579 Missense Mutation TCC,TTC S462F XP_011522888.1
XM_017024424.1 1579 Intron XP_016879913.1
Gene
TAF15
Gene Name
TATA-box binding protein associated factor 15
There are no transcripts associated with this gene.

View Full Product Details