Product Details

SNP ID
rs202032568
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:5033050 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATGGCACCGGCACCAAGCAGGGA[A/G]CCCACTTGGATGGCCACACCAGCTG
Phenotype
MIM: 603060 MIM: 607883
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIF1C PubMed Links

Gene Details

Gene
KIF1C
Gene Name
kinesin family member 1C
There are no transcripts associated with this gene.

Gene
SLC52A1
Gene Name
solute carrier family 52 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001104577.1 1967 Silent Mutation GGC,GGT G418G NP_001098047.1
NM_017986.3 1967 Silent Mutation GGC,GGT G418G NP_060456.3
XM_011523951.1 1967 Silent Mutation GGC,GGT G418G XP_011522253.1

View Full Product Details