Product Details
- SNP ID
-
rs200915259
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:44037493 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GATATGGGGGTGTAATAACAACTTC[C/T]TCCATGACTACGATGTTTTTTTCTT
- Phenotype
-
MIM: 611793
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LSM12
PubMed Links
Gene Details
- Gene
- LSM12
- Gene Name
- LSM12 homolog
View Full Product Details