Product Details

SNP ID
rs201361680
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:14301670 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCTCTATATGTTCCTGTACTCGT[A/G]CGCCGGCTCCTGCGCCGCCGCGCCG
Phenotype
MIM: 604058
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HS3ST3B1 PubMed Links

Gene Details

Gene
HS3ST3B1
Gene Name
heparan sulfate-glucosamine 3-sulfotransferase 3B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006041.2 621 Missense Mutation TAC,TGC Y51C NP_006032.1
XM_017025479.1 621 Intron XP_016880968.1
Gene
MGC12916
Gene Name
uncharacterized protein MGC12916
There are no transcripts associated with this gene.

View Full Product Details