Product Details

SNP ID
rs200162955
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54333649 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTTTGCCTCCTGGCTGCACCTTG[C/G]GGGGCTTCTCTGGCTGTGCTGAGCC
Phenotype
MIM: 607517
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
LILRA4 PubMed Links

Gene Details

Gene
LILRA4
Gene Name
leukocyte immunoglobulin like receptor A4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012276.4 1499 Missense Mutation CCA,GCA P475A NP_036408.4

View Full Product Details