Product Details

SNP ID
rs200164212
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:41332255 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGCGGAAGTCAATGTACAGCTGC[C/T]GCACGCAGCAGTTCTTCTCCGTGGA
Phenotype
MIM: 190180
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC97 PubMed Links

Gene Details

Gene
CCDC97
Gene Name
coiled-coil domain containing 97
There are no transcripts associated with this gene.

Gene
TGFB1
Gene Name
transforming growth factor beta 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011527242.1 1772 Missense Mutation CAG,CGG Q297R XP_011525544.1

View Full Product Details