Product Details
- SNP ID
-
rs200204565
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:19645913 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGAAGGTGGCCATCTGCATGGCCAT[A/G]GCCATGATGTAGACGGTGCTGTTGA
- Phenotype
-
MIM: 609694
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ATP13A1
PubMed Links
Gene Details
- Gene
- ATP13A1
- Gene Name
- ATPase 13A1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_020410.2 |
3349 |
Silent Mutation |
GCC,GCT |
A1107A |
NP_065143.2 |
- Gene
- GMIP
- Gene Name
- GEM interacting protein
There are no transcripts associated with this gene.
View Full Product Details