Product Details

SNP ID
rs199755020
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10554096 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCATCACAGTGGGGCCCAGCAGC[C/T]GTGCCTTCTTGGCCCTCCTCCGCTT
Phenotype
MIM: 611340
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATG4D PubMed Links

Gene Details

Gene
ATG4D
Gene Name
autophagy related 4D cysteine peptidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281504.1 1994 Intron NP_001268433.1
NM_032885.5 1994 Intron NP_116274.3
XM_006722927.1 1994 Intron XP_006722990.1
Gene
KRI1
Gene Name
KRI1 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023008.3 1994 Missense Mutation CAG,CGG Q662R NP_075384.3
XM_011528190.2 1994 Missense Mutation CAG,CGG Q544R XP_011526492.1
Gene
MIR1238
Gene Name
microRNA 1238
There are no transcripts associated with this gene.

View Full Product Details