Product Details

SNP ID
rs199907683
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10554189 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTGTGCTGGGGGACTCTCCGGC[C/T]CCATCAAGCTGCCATCAAGGGCTGG
Phenotype
MIM: 611340
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ATG4D PubMed Links

Gene Details

Gene
ATG4D
Gene Name
autophagy related 4D cysteine peptidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281504.1 1901 Intron NP_001268433.1
NM_032885.5 1901 Intron NP_116274.3
XM_006722927.1 1901 Intron XP_006722990.1
Gene
KRI1
Gene Name
KRI1 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023008.3 1901 Missense Mutation NP_075384.3
XM_011528190.2 1901 Missense Mutation XP_011526492.1
Gene
MIR1238
Gene Name
microRNA 1238
There are no transcripts associated with this gene.

View Full Product Details