Product Details
- SNP ID
-
rs199700435
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:38815898 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCGGCCACCGAATGGTCGCGGGAAT[A/T]CAGCAGGTTGACCTTGGTGCTCTGC
- Phenotype
-
MIM: 600696
MIM: 602518
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ECH1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs58750431] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ECH1
- Gene Name
- enoyl-CoA hydratase 1
- Gene
- LGALS4
- Gene Name
- galectin 4
There are no transcripts associated with this gene.
View Full Product Details