Product Details

SNP ID
rs200854846
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12015445 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCCTTACATTCATACGGCTTCTC[C/T]TCTCTGTGCATCCTTTCATGTATTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101928464 PubMed Links

Gene Details

Gene
LOC101928464
Gene Name
uncharacterized LOC101928464
There are no transcripts associated with this gene.

Gene
ZNF433
Gene Name
zinc finger protein 433
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080411.2 1662 Silent Mutation NP_001073880.1
NM_001308346.1 1662 Silent Mutation NP_001295275.1
NM_001308348.1 1662 Silent Mutation NP_001295277.1
NM_001308351.1 1662 Silent Mutation NP_001295280.1
NM_001308355.1 1662 Silent Mutation NP_001295284.1
NM_001308357.1 1662 Silent Mutation NP_001295286.1
XM_006722661.3 1662 Silent Mutation XP_006722724.1
XM_011527725.2 1662 Silent Mutation XP_011526027.1
XM_011527728.2 1662 Silent Mutation XP_011526030.1
XM_011527729.2 1662 Silent Mutation XP_011526031.1
XM_017026413.1 1662 Intron XP_016881902.1

View Full Product Details