Product Details

SNP ID
rs201196330
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35752210 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCCAAAGCCCGGAACCAACTGGA[G/T]GCTGTCTTGCAGTGTCTGCTGGAGA
Phenotype
MIM: 610695 MIM: 614143 MIM: 607632 MIM: 601080
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HSPB6 PubMed Links

Gene Details

Gene
HSPB6
Gene Name
heat shock protein family B (small) member 6
There are no transcripts associated with this gene.

Gene
IGFLR1
Gene Name
IGF like family receptor 1
There are no transcripts associated with this gene.

Gene
LIN37
Gene Name
lin-37 DREAM MuvB core complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019104.2 433 Missense Mutation GAG,GAT E23D NP_061977.1
Gene
PROSER3
Gene Name
proline and serine rich 3
There are no transcripts associated with this gene.

Gene
PSENEN
Gene Name
presenilin enhancer gamma-secretase subunit
There are no transcripts associated with this gene.

Gene
U2AF1L4
Gene Name
U2 small nuclear RNA auxiliary factor 1 like 4
There are no transcripts associated with this gene.

View Full Product Details