Product Details

SNP ID
rs201101592
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:19645509 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCCAGGCAGAAGTCCAGGAGCAG[C/G]ACCTGGGCAATGACCAGCTTGAACT
Phenotype
MIM: 609694
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ATP13A1 PubMed Links

Gene Details

Gene
ATP13A1
Gene Name
ATPase 13A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020410.2 3556 Silent Mutation GTC,GTG V1176V NP_065143.2
Gene
GMIP
Gene Name
GEM interacting protein
There are no transcripts associated with this gene.

View Full Product Details