Product Details

SNP ID
rs201138591
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:38870737 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCGTGGGGACCCCCAGAATCTGAG[A/G]CGATGCGCACCCGAAGGCTTCGGTA
Phenotype
MIM: 604480
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RINL PubMed Links

Gene Details

Gene
RINL
Gene Name
Ras and Rab interactor like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195833.1 938 Missense Mutation GCC,GTC A286V NP_001182762.1
NM_198445.3 938 Intron NP_940847.1
XM_006723015.2 938 Missense Mutation GCC,GTC A286V XP_006723078.1
XM_006723016.2 938 Missense Mutation GCC,GTC A172V XP_006723079.1
XM_011526454.2 938 Missense Mutation GCC,GTC A286V XP_011524756.1
XM_011526455.2 938 Intron XP_011524757.1
Gene
SIRT2
Gene Name
sirtuin 2
There are no transcripts associated with this gene.

View Full Product Details