Product Details
- SNP ID
-
rs201138737
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:43268146 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGCGGGTTCCAGAAGTTTAAAAGT[C/G]ATGCTAGGAGGGGGAGACAGCATCA
- Phenotype
-
MIM: 176398
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
PSG9
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3746297] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PSG9
- Gene Name
- pregnancy specific beta-1-glycoprotein 9
View Full Product Details