Product Details

SNP ID
rs202007180
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:23654679 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTTTTATGTTCTGCTCTGGCCAA[A/C]ACTCTTGGGCAAAATGAGAACACAT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZNF675 PubMed Links

Gene Details

Gene
ZNF675
Gene Name
zinc finger protein 675
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138330.2 1054 Intron NP_612203.2
XM_017026442.1 1054 Missense Mutation TGT,TTT C9F XP_016881931.1
XM_017026443.1 1054 Missense Mutation TGT,TTT C9F XP_016881932.1
XM_017026444.1 1054 Intron XP_016881933.1

View Full Product Details