Product Details

SNP ID
rs201225377
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:56223726 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCATCTGGTTCACCGAGGAGGCCC[A/G]TTGGCTGGACACGTCTTTCAGATCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZSCAN5A PubMed Links

Gene Details

Gene
ZSCAN5A
Gene Name
zinc finger and SCAN domain containing 5A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322061.1 842 Missense Mutation NP_001308990.1
NM_001322062.1 842 Intron NP_001308991.1
NM_001322064.1 842 Missense Mutation NP_001308993.1
NM_001322065.1 842 Missense Mutation NP_001308994.1
NM_001322066.1 842 Missense Mutation NP_001308995.1
NM_001322067.1 842 Missense Mutation NP_001308996.1
NM_001322068.1 842 Missense Mutation NP_001308997.1
NM_001322069.1 842 Missense Mutation NP_001308998.1
NM_001322070.1 842 Missense Mutation NP_001308999.1
NM_001322072.1 842 Missense Mutation NP_001309001.1
NM_001322073.1 842 Missense Mutation NP_001309002.1
NM_001322074.1 842 Missense Mutation NP_001309003.1
NM_001322075.1 842 Missense Mutation NP_001309004.1
NM_001322076.1 842 Missense Mutation NP_001309005.1
NM_001322077.1 842 Missense Mutation NP_001309006.1
NM_001322078.1 842 Missense Mutation NP_001309007.1
NM_024303.2 842 Intron NP_077279.1
XM_005259254.1 842 Intron XP_005259311.1
XM_006723377.3 842 Missense Mutation XP_006723440.1
XM_011527308.1 842 Intron XP_011525610.1
XM_017027298.1 842 Missense Mutation XP_016882787.1
XM_017027299.1 842 Missense Mutation XP_016882788.1

View Full Product Details