Product Details

SNP ID
rs200977333
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:114675538 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTGTCTCCTACTTACTTTCTCAG[C/G]CTGTTGAGCACCATGATGTTTGCAT
Phenotype
MIM: 102770
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
AMPD1 PubMed Links

Gene Details

Gene
AMPD1
Gene Name
adenosine monophosphate deaminase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000036.2 1818 Missense Mutation AGC,AGG S590R NP_000027.2
NM_001172626.1 1818 Missense Mutation AGC,AGG S586R NP_001166097.1
Gene
DENND2C
Gene Name
DENN domain containing 2C
There are no transcripts associated with this gene.

View Full Product Details