Product Details
- SNP ID
-
rs201193436
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
9
- Location
-
Chr.1:248295484 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTGCTAAACAGGGAGGTCAAAACG[A/G]TGGCCAGAAGCATCATGAAGAGGAC
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
OR2T12
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77223856] are located under a probe and SNP(s) [rs12135684] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR2T12
- Gene Name
- olfactory receptor family 2 subfamily T member 12
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001004692.1 |
95 |
Missense Mutation |
ACC,ATC |
T32I |
NP_001004692.1 |
View Full Product Details