Product Details
- SNP ID
-
rs201099382
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
3
- Location
-
Chr.1:161506420 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCTGTGACTCTGACATGCCAGGGG[G/T]CTCGCAGCCCTGAGAGCGACTCCAT
- Phenotype
-
MIM: 146790
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
FCGR2A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs9427397,rs9427398] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FCGR2A
- Gene Name
- Fc fragment of IgG receptor IIa
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001136219.1 |
231 |
Missense Mutation |
GCT,TCT |
A65S |
NP_001129691.1 |
NM_021642.3 |
231 |
Missense Mutation |
GCT,TCT |
A64S |
NP_067674.2 |
XM_011509287.2 |
231 |
Missense Mutation |
GCT,TCT |
A65S |
XP_011507589.1 |
XM_011509290.2 |
231 |
Missense Mutation |
GCT,TCT |
A65S |
XP_011507592.1 |
XM_011509291.1 |
231 |
Missense Mutation |
GCT,TCT |
A65S |
XP_011507593.1 |
XM_017000663.1 |
231 |
Missense Mutation |
GCT,TCT |
A64S |
XP_016856152.1 |
XM_017000664.1 |
231 |
Missense Mutation |
GCT,TCT |
A65S |
XP_016856153.1 |
XM_017000665.1 |
231 |
Missense Mutation |
GCT,TCT |
A65S |
XP_016856154.1 |
XM_017000666.1 |
231 |
Missense Mutation |
GCT,TCT |
A65S |
XP_016856155.1 |
XM_017000667.1 |
231 |
Missense Mutation |
GCT,TCT |
A65S |
XP_016856156.1 |
XM_017000668.1 |
231 |
Missense Mutation |
GCT,TCT |
A64S |
XP_016856157.1 |
View Full Product Details