Product Details

SNP ID
rs199996657
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:25823669 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCATCCCAATCTGGCAAAACAAGC[C/T]ACATGGGGCTGCTCGAAGTGTAGTA
Phenotype
MIM: 606210
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AUNIP PubMed Links

Gene Details

Gene
AUNIP
Gene Name
aurora kinase A and ninein interacting protein
There are no transcripts associated with this gene.

Gene
LOC646471
Gene Name
uncharacterized LOC646471
There are no transcripts associated with this gene.

Gene
MTFR1L
Gene Name
mitochondrial fission regulator 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099625.1 260 Missense Mutation CCA,CTA P17L NP_001093095.1
NM_001099626.1 260 Missense Mutation CCA,CTA P17L NP_001093096.1
NM_001099627.1 260 Missense Mutation CCA,CTA P17L NP_001093097.1
NM_019557.5 260 Missense Mutation CCA,CTA P17L NP_062457.3
Gene
SEPN1
Gene Name
selenoprotein N, 1
There are no transcripts associated with this gene.

View Full Product Details