Product Details

SNP ID
rs199601281
Assay Type
Functionally Tested
NCBI dbSNP Submissions
3
Location
Chr.1:248239263 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGTGTCCCTGCTTGGAGCTGAAT[A/G]TTTCTTGTTGGCTGTCATGGCTTAT
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
OR2M4 PubMed Links

Gene Details

Gene
OR2M4
Gene Name
olfactory receptor family 2 subfamily M member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017504.1 335 Missense Mutation TAT,TGT Y112C NP_059974.1

View Full Product Details