Product Details

SNP ID
rs199854334
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:27966755 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCTGCTCTGGGTCTGGCTTCAGG[A/G]CACAGACTTCATGCCGGACCCCAGC
Phenotype
MIM: 601655
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EYA3 PubMed Links

Gene Details

Gene
EYA3
Gene Name
EYA transcriptional coactivator and phosphatase 3
There are no transcripts associated with this gene.

Gene
SMPDL3B
Gene Name
sphingomyelin phosphodiesterase acid like 3B
There are no transcripts associated with this gene.

Gene
XKR8
Gene Name
XK related 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018053.3 1351 Missense Mutation GAC,GGC D248G NP_060523.2
XM_011541679.2 1351 Missense Mutation GAC,GGC D302G XP_011539981.1
XM_011541680.2 1351 Missense Mutation GAC,GGC D266G XP_011539982.1

View Full Product Details