Product Details
- SNP ID
-
rs199524870
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
5
- Location
-
Chr.1:151294436 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCACAAACTCTGTGGTCAGCTTAAA[A/G]GCTGACGTCTCAAAGCCCAGATTTC
- Phenotype
-
MIM: 602758
MIM: 610568
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PI4KB
PubMed Links
Gene Details
- Gene
- PI4KB
- Gene Name
- phosphatidylinositol 4-kinase beta
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001198774.1 |
2549 |
Silent Mutation |
GCC,GCT |
A692A |
NP_001185703.1 |
XM_005245264.3 |
2549 |
Silent Mutation |
GCC,GCT |
A704A |
XP_005245321.1 |
XM_011509634.1 |
2549 |
Silent Mutation |
GCC,GCT |
A719A |
XP_011507936.1 |
XM_011509635.1 |
2549 |
Silent Mutation |
GCC,GCT |
A707A |
XP_011507937.1 |
XM_011509636.2 |
2549 |
Silent Mutation |
GCC,GCT |
A707A |
XP_011507938.1 |
XM_011509637.2 |
2549 |
Silent Mutation |
GCC,GCT |
A707A |
XP_011507939.1 |
XM_011509638.1 |
2549 |
Silent Mutation |
GCC,GCT |
A704A |
XP_011507940.1 |
XM_017001486.1 |
2549 |
Silent Mutation |
GCC,GCT |
A692A |
XP_016856975.1 |
XM_017001487.1 |
2549 |
Silent Mutation |
GCC,GCT |
A692A |
XP_016856976.1 |
XM_017001488.1 |
2549 |
Silent Mutation |
GCC,GCT |
A692A |
XP_016856977.1 |
XM_017001489.1 |
2549 |
Silent Mutation |
GCC,GCT |
A707A |
XP_016856978.1 |
XM_017001490.1 |
2549 |
Silent Mutation |
GCC,GCT |
A707A |
XP_016856979.1 |
XM_017001491.1 |
2549 |
Silent Mutation |
GCC,GCT |
A707A |
XP_016856980.1 |
XM_017001492.1 |
2549 |
Silent Mutation |
GCC,GCT |
A692A |
XP_016856981.1 |
- Gene
- ZNF687
- Gene Name
- zinc finger protein 687
There are no transcripts associated with this gene.
View Full Product Details