Product Details

SNP ID
rs200019982
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:162399064 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGTTCCTTTAGGCTTGGAAAGAC[A/C]TGTTTTGGAGAACCTTCTGCAGTCT
Phenotype
MIM: 608510
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SH2D1B PubMed Links

Gene Details

Gene
SH2D1B
Gene Name
SH2 domain containing 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_053282.4 344 Missense Mutation CAG,CAT Q74H NP_444512.2

View Full Product Details