Product Details

SNP ID
rs200155577
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:27347548 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGACCTTGTCCGAGCGTCTATG[C/T]GCAGGAAGAAGAGCACCAGGGGTGA
Phenotype
MIM: 604468 MIM: 608042
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAP3K6 PubMed Links

Gene Details

Gene
MAP3K6
Gene Name
mitogen-activated protein kinase kinase kinase 6
There are no transcripts associated with this gene.

Gene
SYTL1
Gene Name
synaptotagmin like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193308.1 516 Missense Mutation CGC,TGC R107C NP_001180237.1
NM_032872.2 516 Missense Mutation CGC,TGC R107C NP_116261.1
XM_005246022.3 516 Missense Mutation CGC,TGC R139C XP_005246079.1
XM_006710990.1 516 Missense Mutation CGC,TGC R139C XP_006711053.1
XM_006710991.1 516 UTR 5 XP_006711054.1

View Full Product Details