Product Details

SNP ID
rs200877537
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:3021747 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTTTTGACAATGGCTCGGGGTTC[C/T]GCAAAGCGGGCCTGTCTGGGGAGTT
Phenotype
MIM: 608535
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACTRT2 PubMed Links

Gene Details

Gene
ACTRT2
Gene Name
actin related protein T2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080431.4 266 Missense Mutation CGC,TGC R21C NP_536356.3

View Full Product Details