Product Details

SNP ID
rs200493380
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:152698986 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCACCCAAGTGTCCTCCAAAGTGC[C/T]GACCTCAGTGCCCAGCCCCATGCCC
Phenotype
MIM: 612609
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LCE2A PubMed Links

Gene Details

Gene
LCE2A
Gene Name
late cornified envelope 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178428.3 136 Nonsense Mutation CGA,TGA R29* NP_848515.1

View Full Product Details