Product Details

SNP ID
rs200520550
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:152699090 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCTGGGGGCTGCTGCAGCTCTGG[A/G]GGTGGCGGCTGCTGCCTGAGCCACC
Phenotype
MIM: 612609 MIM: 612618
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LCE2A PubMed Links

Gene Details

Gene
LCE2A
Gene Name
late cornified envelope 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178428.3 240 Silent Mutation GGA,GGG G63G NP_848515.1
Gene
LCE4A
Gene Name
late cornified envelope 4A
There are no transcripts associated with this gene.

View Full Product Details