Product Details

SNP ID
rs200940187
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:53459511 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCGAGATGCAGGAACCATGGCTTC[C/T]TGGTGCCCGTCAAGGGACACGCGGG
Phenotype
MIM: 614805
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DMRTB1 PubMed Links

Gene Details

Gene
DMRTB1
Gene Name
DMRT like family B with proline rich C-terminal 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033067.2 113 Silent Mutation CTG,TTG L20L NP_149056.1

View Full Product Details