Product Details

SNP ID
rs201272727
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:25417181 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGACACTGTTCTCTGTTAAGAAATC[A/G]ACGCTGCACTGTAGCATACCTGTAA
Phenotype
MIM: 610608
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
GINS1 PubMed Links

Gene Details

Gene
GINS1
Gene Name
GINS complex subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021067.4 313 Missense Mutation CAA,CGA Q73R NP_066545.3
XM_017028162.1 313 Missense Mutation CAA,CGA Q38R XP_016883651.1

View Full Product Details