Product Details

SNP ID
rs200268997
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:33237748 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGGGCCTCATTGTCTTCTACGGG[C/T]TGTTAGCCCAGACCATGGCCCAGTT
Phenotype
MIM: 607412
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BPIFA1 PubMed Links

Gene Details

Gene
BPIFA1
Gene Name
BPI fold containing family A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243193.1 107 Silent Mutation CTG,TTG L13L NP_001230122.1
NM_016583.3 107 Silent Mutation CTG,TTG L13L NP_057667.1
NM_130852.2 107 Silent Mutation CTG,TTG L13L NP_570913.1
Gene
BPIFA3
Gene Name
BPI fold containing family A member 3
There are no transcripts associated with this gene.

View Full Product Details