Product Details

SNP ID
rs200422001
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:388039 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAGCCACCCCTCTGGCTGCTCCTG[C/T]GGGTTCCCTGTCCAGGAAGAAGCGG
Phenotype
MIM: 607898
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRIB3 PubMed Links

Gene Details

Gene
TRIB3
Gene Name
tribbles pseudokinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301188.1 78 Missense Mutation GCG,GTG A10V NP_001288117.1
NM_001301190.1 78 Missense Mutation GCG,GTG A10V NP_001288119.1
NM_001301193.1 78 Missense Mutation GCG,GTG A10V NP_001288122.1
NM_001301196.1 78 Missense Mutation GCG,GTG A10V NP_001288125.1
NM_001301201.1 78 Missense Mutation GCG,GTG A37V NP_001288130.1
NM_021158.4 78 Missense Mutation GCG,GTG A10V NP_066981.2
XM_017027989.1 78 Missense Mutation GCG,GTG A37V XP_016883478.1

View Full Product Details