Product Details

SNP ID
rs200286160
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:34715323 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTATCAAGTCGCAGTCCTGCTTGTT[A/T]ACTTGGATTTTCTTCGCTTGGATTG
Phenotype
MIM: 605299
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
NCOA6 PubMed Links

Gene Details

Gene
NCOA6
Gene Name
nuclear receptor coactivator 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_006723755.3 5264 Intron XP_006723818.1
XM_017027743.1 5264 Silent Mutation GTA,GTT V2038V XP_016883232.1
XM_017027746.1 5264 Intron XP_016883235.1
XM_017027748.1 5264 UTR 3 XP_016883237.1
XM_017027749.1 5264 Intron XP_016883238.1
Gene
TP53INP2
Gene Name
tumor protein p53 inducible nuclear protein 2
There are no transcripts associated with this gene.

View Full Product Details