Product Details

SNP ID
rs201505970
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45824067 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCGATCTCCTCGTCCGTCACGTG[C/T]TCCCCGGAGGCCCTGAAAATCTCAG
Phenotype
MIM: 191039 MIM: 605574
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SNX21 PubMed Links

Gene Details

Gene
SNX21
Gene Name
sorting nexin family member 21
There are no transcripts associated with this gene.

Gene
TNNC2
Gene Name
troponin C2, fast skeletal type
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003279.2 451 Silent Mutation GAA,GAG E125E NP_003270.1
XM_011529031.2 451 Silent Mutation GAA,GAG E110E XP_011527333.1
Gene
UBE2C
Gene Name
ubiquitin conjugating enzyme E2 C
There are no transcripts associated with this gene.

View Full Product Details