Product Details
- SNP ID
-
rs200398348
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:23568317 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAACTCCACTGTGGCAGGGAGGTAA[C/G]GAGCCATGACATTGTGTTCATCACA
- Phenotype
-
MIM: 616536
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CST9L
PubMed Links
Gene Details
- Gene
- CST9L
- Gene Name
- cystatin 9-like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_080610.2 |
433 |
Missense Mutation |
CCT,CGT |
P45R |
NP_542177.1 |
View Full Product Details