Product Details

SNP ID
rs201413966
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:663779 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTGAGCGCGAAGCTCTTGTCGCA[C/G]TGGCGGCAGCGGTAGTGCTTGAAGG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SCRT2 PubMed Links

Gene Details

Gene
SCRT2
Gene Name
scratch family transcriptional repressor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033129.3 1394 Missense Mutation CAC,CAG H272Q NP_149120.1

View Full Product Details