Product Details

SNP ID
rs200490066
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:32150395 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTGATGTCATGAAGTCTTCAAGA[C/G]AGACAGATAGGCTTTCACTGAGATC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C22orf42 PubMed Links

Gene Details

Gene
C22orf42
Gene Name
chromosome 22 open reading frame 42
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010859.1 1028 Missense Mutation TCT,TGT S193C NP_001010859.1
XM_011529922.2 1028 Intron XP_011528224.1
XM_017028629.1 1028 Intron XP_016884118.1
XM_017028630.1 1028 Missense Mutation TCT,TGT S176C XP_016884119.1
XM_017028631.1 1028 UTR 3 XP_016884120.1
XM_017028632.1 1028 Intron XP_016884121.1

View Full Product Details