Product Details

SNP ID
rs200910750
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:41998870 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACCGCCGCGGAGCCCTCATCCCT[A/G]ACGGTGAAAGGTGCCTGAGGGGAAG
Phenotype
MIM: 608314 MIM: 610981
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SEPT3 PubMed Links

Gene Details

Gene
SEPT3
Gene Name
septin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019106.5 146 Intron NP_061979.3
NM_145733.2 146 Intron NP_663786.2
XM_017028862.1 146 Intron XP_016884351.1
Gene
WBP2NL
Gene Name
WBP2 N-terminal like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152613.2 146 Missense Mutation AAC,GAC N18D NP_689826.2

View Full Product Details