Product Details

SNP ID
rs200757651
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:233637141 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATGCCATGACTTTCAAGGAGAGA[A/G]TATGGAACCACATCGTGCACTTGGA
Phenotype
MIM: 606435 MIM: 606433
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UGT1A10 PubMed Links

Gene Details

Gene
UGT1A10
Gene Name
UDP glucuronosyltransferase family 1 member A10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019075.2 665 Missense Mutation ATA,GTA I207V NP_061948.1
Gene
UGT1A8
Gene Name
UDP glucuronosyltransferase family 1 member A8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019076.4 665 Intron NP_061949.3

View Full Product Details