Product Details

SNP ID
rs200526441
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:88028140 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATGAGTACTGGTACTGTTATCTG[A/G]GGAGAAGTGCTTGTAAACTCTATGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRCC1 PubMed Links

Gene Details

Gene
KRCC1
Gene Name
lysine rich coiled-coil 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304526.1 1191 Missense Mutation CCA,TCA P142S NP_001291455.1
NM_016618.2 1191 Missense Mutation CCA,TCA P142S NP_057702.1
XM_017004292.1 1191 Missense Mutation CCA,TCA P142S XP_016859781.1

View Full Product Details