Product Details
- SNP ID
-
rs199693434
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:178460396 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAGGAGGATTTGAAAGGGAAGAAA[C/T]GGCAACATTTGCACTGCTGTACAGG
- Phenotype
-
MIM: 610219
MIM: 607062
MIM: 603424
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DFNB59
PubMed Links
Gene Details
- Gene
- DFNB59
- Gene Name
- deafness, autosomal recessive 59
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001042702.3 |
1443 |
Missense Mutation |
ACG,ATG |
T239M |
NP_001036167.1 |
XM_005246627.2 |
1443 |
Missense Mutation |
ACG,ATG |
T242M |
XP_005246684.1 |
XM_005246628.3 |
1443 |
Intron |
|
|
XP_005246685.1 |
XM_005246629.3 |
1443 |
Missense Mutation |
ACG,ATG |
T236M |
XP_005246686.1 |
XM_011511247.2 |
1443 |
Missense Mutation |
ACG,ATG |
T274M |
XP_011509549.1 |
XM_011511249.2 |
1443 |
Missense Mutation |
ACG,ATG |
T80M |
XP_011509551.1 |
XM_011511250.2 |
1443 |
Missense Mutation |
ACG,ATG |
T80M |
XP_011509552.1 |
XM_011511251.1 |
1443 |
Missense Mutation |
ACG,ATG |
T80M |
XP_011509553.1 |
XM_017004221.1 |
1443 |
Missense Mutation |
ACG,ATG |
T274M |
XP_016859710.1 |
XM_017004222.1 |
1443 |
Missense Mutation |
ACG,ATG |
T80M |
XP_016859711.1 |
XM_017004223.1 |
1443 |
Missense Mutation |
ACG,ATG |
T80M |
XP_016859712.1 |
XM_017004224.1 |
1443 |
Missense Mutation |
ACG,ATG |
T80M |
XP_016859713.1 |
XM_017004225.1 |
1443 |
Missense Mutation |
ACG,ATG |
T80M |
XP_016859714.1 |
- Gene
- FKBP7
- Gene Name
- FK506 binding protein 7
There are no transcripts associated with this gene.
- Gene
- PRKRA
- Gene Name
- protein activator of interferon induced protein kinase EIF2AK2
There are no transcripts associated with this gene.
View Full Product Details