Product Details

SNP ID
rs200156014
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:160277367 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCAAGTAGGCTCCTTGCATAGCT[G/T]ACGTTGCAGGCATGTACTAGAAAGA
Phenotype
MIM: 602310
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC100505984 PubMed Links

Gene Details

Gene
LOC100505984
Gene Name
uncharacterized LOC100505984
There are no transcripts associated with this gene.

Gene
RBMS1
Gene Name
RNA binding motif single stranded interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002897.4 1487 Nonsense Mutation TAA,TCA *357S NP_002888.1
NM_016836.3 1487 Nonsense Mutation TAA,TCA *360S NP_058520.1
XM_005246737.1 1487 Nonsense Mutation TAA,TCA *376S XP_005246794.1
XM_005246738.2 1487 Nonsense Mutation TAA,TCA *375S XP_005246795.1
XM_005246739.2 1487 Nonsense Mutation TAA,TCA *374S XP_005246796.1
XM_005246740.1 1487 Nonsense Mutation TAA,TCA *373S XP_005246797.1
XM_005246741.3 1487 Nonsense Mutation TAA,TCA *373S XP_005246798.1
XM_017004626.1 1487 Nonsense Mutation TAA,TCA *356S XP_016860115.1

View Full Product Details